Assay Details
| Gene Symbol: Gcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017186.2 | NP_058882.1 | 3 - 4 | 483 | 89 | |
| GenBank mRNA | AF081557.1 | - | 3 - 4 | 484 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gcm1
Genomic Map