Assay Details
| Gene Symbol: Gcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017186.2 | NP_058882.1 | 4 - 5 | 600 | 59 | |
| GenBank mRNA | AF081557.1 | - | 4 - 5 | 601 | 59 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gcm1
Genomic Map