Assay Details
| Gene Symbol: Ubl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048243.2 | NP_001041708.2 | 2 - 3 | 175 | 62 | |
| XM_006242661.2 | XP_006242723.1 | 2 - 3 | 356 | 62 | ||
| GenBank mRNA | BC091358.1 | - | 1 - 2 | 369 | 62 | |
| BC157818.1 | - | 1 - 2 | 78 | 7936655 | 62 | |
| CB713235.1 | - | 2 - 3 | 175 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ubl5
Genomic Map