Assay Details
| Gene Symbol: Ubl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048243.2 | NP_001041708.2 | 3 - 4 | 262 | 86 | |
| XM_006242661.2 | XP_006242723.1 | 3 - 4 | 443 | 86 | ||
| GenBank mRNA | BC157818.1 | - | 2 - 3 | 165 | 7936655 | 86 |
| CB713235.1 | - | 3 - 4 | 262 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ubl5
Genomic Map