Assay Details
| Gene Symbol: Zfyve28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107227.1 | NP_001100697.1 | 4 - 5 | 690 | 73 | |
| XM_006251256.2 | XP_006251318.1 | 4 - 5 | 780 | 73 | ||
| XM_006251257.2 | XP_006251319.1 | 4 - 5 | 780 | 73 | ||
| XM_006251258.2 | XP_006251320.1 | 4 - 5 | 781 | 73 | ||
| XM_006251259.2 | XP_006251321.1 | 4 - 5 | 776 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfyve28
Genomic Map