Assay Details
| Gene Symbol: Morc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106016.1 | NP_001099486.1 | 7 - 8 | 703 | 73 | |
| XM_006251198.2 | XP_006251260.1 | 9 - 10 | 846 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Morc2
Genomic Map