Assay Details
| Gene Symbol: LOC100910240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250645.2 | XP_006250707.1 | 951 | 117 | ||
| Gene Symbol: Ptpn13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100789.1 | NP_001094259.1 | 34 - 35 | 5937 | 117 | |
| XM_006250672.2 | XP_006250734.1 | 34 - 35 | 5967 | 117 | ||
| XM_006250673.2 | XP_006250735.1 | 33 - 34 | 5421 | 117 | ||
| XM_006250674.2 | XP_006250736.1 | 31 - 32 | 5385 | 117 | ||
| XM_008770051.1 | XP_008768273.1 | 31 - 32 | 5400 | 117 | ||
| XM_008770052.1 | XP_008768274.1 | 34 - 35 | 5967 | 117 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910240
Genomic Map