Assay Details
| Gene Symbol: Vstm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029920.1 | NP_001025091.1 | 7 - 8 | 863 | 63 | |
| GenBank mRNA | BC098063.1 | - | 7 - 8 | 863 | 7458419 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P803941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56637 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P803941 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P803941 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P803941 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P803941 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41187 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73358 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28071 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67914 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10786841 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786837 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786845 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786840 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786844 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786843 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786842 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786839 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786838 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10786836 | - | - |
| Rat Genome 230 2.0 Array | 1373596_at | - | - |
| Rat Genome 230 2.0 Array | 1373596_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373596_at | - | - |
| Affymetrix HT Rat Focus Array | 1373596_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600001424.rn.1 | TC1600000124.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600001425.rn.1 | TC1600000124.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373596_at | - | - |
| Rat Expression Set 230 | 1373596_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10786836 | - | - |
| Rat Genome U34 Set | rc_AA956769_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21849467 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371902 | - | - |

Vstm4
Genomic Map