Assay Details
| Gene Symbol: Tmem110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198774.2 | NP_942069.1 | 3 - 4 | 391 | 65 | |
| GenBank mRNA | AY283801.1 | - | 3 - 4 | 414 | 65 | |
| BC088092.1 | - | 3 - 4 | 391 | 7368117 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem110
Genomic Map