Assay Details
| Gene Symbol: Prl2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053791.2 | NP_446243.1 | 2 - 3 | 252 | 141 | |
| GenBank mRNA | AF226608.1 | - | 2 - 3 | 250 | 141 | |
| BC097296.1 | - | 2 - 3 | 252 | 7445776 | 141 | |
| DQ329279.1 | - | 2 - 3 | 250 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prl2a1
Genomic Map