Assay Details
| Gene Symbol: Nol7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253849.2 | XP_006253911.1 | 3 - 4 | 518 | 112 | |
| GenBank mRNA | FQ210031.1 | - | 3 - 4 | 390 | 112 | |
| FQ227425.1 | - | 3 - 4 | 386 | 112 | ||
| FQ229485.1 | - | 3 - 4 | 400 | 112 | ||
More Information
Set Membership: |
|

Nol7
Genomic Map