Assay Details
| Gene Symbol: Spata6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014165.1 | NP_001014187.1 | 2 - 3 | 292 | 75 | |
| XM_006231237.2 | XP_006231299.1 | 2 - 3 | 251 | 75 | ||
| XM_006231239.2 | XP_006231301.1 | 2 - 3 | 168 | 75 | ||
| XM_006231240.2 | XP_006231302.1 | 1 - 2 | 138 | 75 | ||
| XM_008760342.1 | XP_008758564.1 | 3 - 4 | 321 | 75 | ||
| XM_008760343.1 | XP_008758565.1 | 3 - 4 | 322 | 75 | ||
| GenBank mRNA | BC079024.1 | - | 2 - 3 | 292 | 7113918 | 75 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006231239.2, XM_006231240.2 transcripts.
Set Membership: |
|

Spata6l
Genomic Map