Assay Details
| Gene Symbol: Trim46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107691.1 | NP_001101161.1 | 3 - 4 | 694 | 69 | |
| XM_008761170.1 | XP_008759392.1 | 3 - 4 | 811 | 69 | ||
| XM_008761171.1 | XP_008759393.1 | 3 - 4 | 658 | 69 | ||
| XM_008761172.1 | XP_008759394.1 | 3 - 4 | 657 | 69 | ||
| XM_008761173.1 | XP_008759395.1 | 3 - 4 | 658 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim46
Genomic Map