Assay Details
| Gene Symbol: Paqr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191077.1 | NP_001178006.1 | 1 - 2 | 37 | 51 | |
| XM_006232755.2 | XP_006232817.1 | 1 - 2 | 1039 | 51 | ||
| XM_008761210.1 | XP_008759432.1 | 1 - 2 | 1039 | 51 | ||
| XM_008761211.1 | XP_008759433.1 | 1 - 2 | 1039 | 51 | ||
| XM_008761213.1 | XP_008759435.1 | 1 - 2 | 1039 | 51 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191077.1 transcripts.
Set Membership: |
|

Paqr6
Genomic Map