Assay Details
| Gene Symbol: Paqr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191077.1 | NP_001178006.1 | 7 - 8 | 819 | 88 | |
| XM_006232755.2 | XP_006232817.1 | 7 - 8 | 1821 | 88 | ||
| XM_008761210.1 | XP_008759432.1 | 6 - 7 | 1896 | 88 | ||
| XM_008761211.1 | XP_008759433.1 | 6 - 7 | 1764 | 88 | ||
| XM_008761212.1 | XP_008759434.1 | 6 - 7 | 1001 | 88 | ||
| XM_008761213.1 | XP_008759435.1 | 5 - 6 | 1491 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Paqr6
Genomic Map