Assay Details
| Gene Symbol: Dohh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025006.1 | NP_001020177.1 | 1 - 2 | 71 | 61 | |
| GenBank mRNA | BC087658.1 | - | 1 - 2 | 71 | 7314221 | 61 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC087658.1, NM_001025006.1 transcripts.
Set Membership: |
|

Dohh
Genomic Map