Assay Details
| Gene Symbol: Tmem106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024967.1 | NP_001020138.1 | 4 - 5 | 728 | 71 | |
| XM_008767977.1 | XP_008766199.1 | 4 - 5 | 926 | 71 | ||
| XM_008767978.1 | XP_008766200.1 | 5 - 6 | 669 | 71 | ||
| XM_008767979.1 | XP_008766201.1 | 5 - 6 | 657 | 71 | ||
| GenBank mRNA | BC091170.1 | - | 4 - 5 | 728 | 7131003 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem106a
Genomic Map