Assay Details
| Gene Symbol: Oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107295.1 | NP_001100765.1 | 5 - 6 | 638 | 61 | |
| XM_006252623.2 | XP_006252685.1 | 5 - 6 | 739 | 61 | ||
| XM_006252624.2 | XP_006252686.1 | 6 - 7 | 846 | 61 | ||
| XM_006252625.2 | XP_006252687.1 | 6 - 7 | 832 | 61 | ||
| XM_006252626.2 | XP_006252688.1 | 5 - 6 | 725 | 61 | ||
| XM_008771000.1 | XP_008769222.1 | 6 - 7 | 927 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oxnad1
Genomic Map