Assay Details
| Gene Symbol: Pwwp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108507.2 | NP_001101977.1 | 1 - 2 | 154 | 72 | |
| XM_006230466.2 | XP_006230528.1 | 1 - 2 | 2523 | 72 | ||
| XM_006230469.2 | XP_006230531.1 | 1 - 2 | 2522 | 72 | ||
| GenBank mRNA | BC162015.1 | - | 1 - 2 | 154 | 9092167 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

Pwwp2b
Genomic Map