Assay Details
| Gene Symbol: March5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106372.2 | NP_001099842.1 | 1 - 2 | 326 | 61 | |
| XM_008760367.1 | XP_008758589.1 | 1 - 2 | 332 | 61 | ||
| GenBank mRNA | BC158801.1 | - | 1 - 2 | 326 | 7321043 | 61 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

March5
Genomic Map