Assay Details
| Gene Symbol: Rras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108481.1 | NP_001101951.1 | 2 - 3 | 251 | 86 | |
| GenBank mRNA | FQ211166.1 | - | 2 - 3 | 267 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rras
Genomic Map