Assay Details
| Gene Symbol: Map9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135716.1 | NP_001129188.1 | 7 - 8 | 1251 | 108 | |
| XM_006232515.2 | XP_006232577.1 | 7 - 8 | 1313 | 108 | ||
| XM_006232516.2 | XP_006232578.1 | 6 - 7 | 1165 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map9
Genomic Map