Assay Details
| Gene Symbol: Cttnbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114401.1 | NP_001107873.1 | 8 - 9 | 2834 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cttnbp2| Gene Symbol: Cttnbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114401.1 | NP_001107873.1 | 8 - 9 | 2834 | 64 | |
Set Membership: |
|