Assay Details
| Gene Symbol: Arf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024149.1 | NP_077063.1 | 3 - 4 | 351 | 58 | |
| GenBank mRNA | BC087692.1 | - | 3 - 4 | 313 | 7310424 | 58 |
| L12384.1 | - | 3 - 4 | 351 | 58 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arf5
Genomic Map