Assay Details
| Gene Symbol: Slc16a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105797.2 | NP_001099267.2 | 5 - 6 | 737 | 102 | |
| XM_006246646.1 | XP_006246708.1 | 5 - 6 | 877 | 102 | ||
| XM_006246647.2 | XP_006246709.1 | 4 - 5 | 1420 | 102 | ||
| XM_006246648.2 | XP_006246710.1 | 4 - 5 | 722 | 102 | ||
| XM_006246649.2 | XP_006246711.1 | 3 - 4 | 441 | 102 | ||
| XM_008767805.1 | XP_008766027.1 | 4 - 5 | 1281 | 102 | ||
| GenBank mRNA | BC169003.1 | - | 5 - 6 | 877 | 7122746 | 102 |
| FQ219143.1 | - | 2 - 3 | 212 | 102 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc16a11
Genomic Map