Assay Details
| Gene Symbol: Csf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053852.1 | NP_446304.1 | 3 - 4 | 325 | 127 | |
| GenBank mRNA | U00620.1 | - | 3 - 4 | 274 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csf2
Genomic Map