Assay Details
| Gene Symbol: Adad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006977.2 | NP_001006978.2 | 9 - 10 | 1387 | 80 | |
| XM_006232278.2 | XP_006232340.1 | 9 - 10 | 1571 | 80 | ||
| XM_006232279.2 | XP_006232341.1 | 10 - 11 | 1423 | 80 | ||
| GenBank mRNA | BC105906.1 | - | 9 - 10 | 1453 | 7453283 | 80 |
More Information
Updated Mapping Information
Set Membership: |
|

Adad1
Genomic Map