Assay Details
| Gene Symbol: Gtf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113570.1 | NP_001107042.1 | 4 - 5 | 636 | 98 | |
| GenBank mRNA | AF391798.1 | - | 4 - 5 | 397 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf3a
Genomic Map