Assay Details
| Gene Symbol: Ccz1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014126.1 | NP_001014148.1 | 12 - 13 | 2218 | 119 | |
| XM_006248878.1 | XP_006248940.1 | 13 - 14 | 1341 | 119 | ||
| GenBank mRNA | BC079086.1 | - | 12 - 13 | 2218 | 7123188 | 119 |
More Information
Updated Mapping Information
Set Membership: |
|

Ccz1b
Genomic Map