Assay Details
| Gene Symbol: Ccz1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014126.1 | NP_001014148.1 | 4 - 5 | 458 | 149 | |
| XM_006248878.1 | XP_006248940.1 | 4 - 5 | 468 | 149 | ||
| XM_006248879.1 | XP_006248941.1 | 4 - 5 | 468 | 149 | ||
| GenBank mRNA | BC079086.1 | - | 4 - 5 | 458 | 7123188 | 149 |
More Information
Updated Mapping Information
Set Membership: |
|

Ccz1b
Genomic Map