Assay Details
| Gene Symbol: Rbmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113786.2 | NP_001107258.2 | 3 - 4 | 311 | 80 | |
| GenBank mRNA | BC158684.1 | - | 3 - 4 | 311 | 8373764 | 80 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbmx2
Genomic Map