Assay Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001053214.4 | XP_001053214.2 | 10 - 11 | 1409 | 64 | |
| XM_006228482.1 | XP_006228544.1 | 10 - 11 | 1581 | 64 | ||
| XM_006228483.2 | XP_006228545.1 | 11 - 12 | 2118 | 64 | ||
| XM_006228484.2 | XP_006228546.1 | 11 - 12 | 1472 | 64 | ||
| XM_006228485.2 | XP_006228547.1 | 11 - 12 | 1801 | 64 | ||
| XM_006228486.2 | XP_006228548.1 | 10 - 11 | 1973 | 64 | ||
| XM_006228487.1 | XP_006228549.1 | 10 - 11 | 1332 | 64 | ||
| XM_006228488.2 | XP_006228550.1 | 11 - 12 | 1684 | 64 | ||
| XM_006228489.1 | XP_006228551.1 | 11 - 12 | 1471 | 64 | ||
| XM_006228491.2 | XP_006228553.1 | 11 - 12 | 1595 | 64 | ||
| XM_006228493.1 | XP_006228555.1 | 10 - 11 | 1581 | 64 | ||
| XM_008759031.1 | XP_008757253.1 | 10 - 11 | 1526 | 64 | ||
More Information
Set Membership: |
|

Zc3h4
Genomic Map