Assay Details
| Gene Symbol: Ppp1r15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133546.3 | NP_598230.2 | 1 - 2 | 218 | 115 | |
| XM_006228974.2 | XP_006229036.1 | 1 - 2 | 280 | 115 | ||
| GenBank mRNA | BC072513.1 | - | 1 - 2 | 200 | 7104357 | 115 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC072513.1, NM_133546.3, XM_006228974.2 transcripts.
Set Membership: |
|

Ppp1r15a
Genomic Map