Assay Details
| Gene Symbol: Zfp180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271280.1 | NP_001258209.1 | 2 - 3 | 423 | 92 | |
| NM_144757.3 | NP_653358.2 | 2 - 3 | 341 | 92 | ||
| XM_006228386.2 | XP_006228448.1 | 2 - 3 | 374 | 92 | ||
| XM_006228387.2 | XP_006228449.1 | 2 - 3 | 425 | 92 | ||
| GenBank mRNA | U41164.1 | - | 2 - 3 | 341 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271280.1, U41164.1, XM_006228387.2 transcripts.
Set Membership: |
|

Zfp180
Genomic Map