Assay Details
| Gene Symbol: Nmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107682.2 | NP_001101152.1 | 10 - 11 | 1017 | 74 | |
| XM_008761090.1 | XP_008759312.1 | 10 - 11 | 1012 | 74 | ||
| GenBank mRNA | BC161998.1 | - | 10 - 11 | 1017 | 9035992 | 74 |
| FQ215207.1 | - | 8 - 9 | 754 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nmd3
Genomic Map