Assay Details
| Gene Symbol: Timp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109393.1 | NP_001102863.1 | 2 - 3 | 607 | 126 | |
| GenBank mRNA | FQ213131.1 | - | 2 - 3 | 344 | 126 | |
| FQ214060.1 | - | 2 - 3 | 315 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timp4
Genomic Map