Assay Details
| Gene Symbol: Mbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001059437.5 | XP_001059437.3 | 6 - 7 | 1676 | 89 | |
| GenBank mRNA | BC158892.1 | - | 6 - 7 | 1016 | 89 | |
More Information
Set Membership: |
|

Mbd4
Genomic Map