Assay Details
| Gene Symbol: Iqsec3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207617.1 | NP_997500.1 | 6 - 7 | 2923 | 87 | |
| GenBank mRNA | AB057643.1 | - | 6 - 7 | 2923 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P037666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19591 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P037666 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P037666 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P037666 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P190381 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P172682 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102077 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36328 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44296 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57625 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55473 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53489 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30305 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5648 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55595 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10865279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865284 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865285 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865283 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865281 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865282 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10865270 | - | - |
| Rat Genome 230 2.0 Array | 1391036_at | - | - |
| Rat Genome 230 2.0 Array | 1391036_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391036_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391036_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400025904.rn.1 | TC0400003678.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400025913.rn.1 | TC0400003678.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400025907.rn.1 | TC0400003678.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400025908.rn.1 | TC0400003678.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400025905.rn.1 | TC0400003678.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400025906.rn.1 | TC0400003678.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400025912.rn.1 | TC0400003678.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400025909.rn.1 | TC0400003678.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400025911.rn.1 | TC0400003678.rn.1 | RTA TC |
| Rat Expression Set 230 | 1391036_PM_at | - | - |
| Rat Expression Set 230 | 1391036_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10865270 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21825344 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355804 | - | - |
| RatRef-12 Expression BeadChip | ILMN_55068 | - | - |

Iqsec3
Genomic Map