Assay Details
| Gene Symbol: Far2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237697.2 | XP_006237759.1 | 8 - 9 | 2062 | 64 | |
| XM_575726.6 | XP_575726.2 | 8 - 9 | 1359 | 64 | ||
More Information
Set Membership: |
|

Far2
Genomic Map