Assay Details
| Gene Symbol: Map3k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107058.2 | NP_001100528.1 | 6 - 7 | 793 | 56 | |
| XM_008768358.1 | XP_008766580.1 | 6 - 7 | 651 | 56 | ||
| GenBank mRNA | BC168979.1 | - | 6 - 7 | 793 | 7125270 | 56 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
blood vessel development
biological_process
intracellular signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
protein autophosphorylation
positive regulation of p38MAPK cascade
molecular_function
protein kinase activity
MAP kinase kinase activity
ATP binding
MAPK cascade
activation of MAPK activity
blood vessel development
biological_process
intracellular signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
protein autophosphorylation
positive regulation of p38MAPK cascade
molecular_function
protein kinase activity
MAP kinase kinase activity
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P321669 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P10970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65080 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P321669 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10970 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10970 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P321669 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P321669 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P10970 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P321669 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10970 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P321675 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18542 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62339 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10739111 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739109 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739113 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739107 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739112 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739104 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739103 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739116 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739105 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739114 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739106 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739115 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739101 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739110 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739100 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739108 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10739099 | - | - |
| Rat Genome 230 2.0 Array | 1385814_at | - | - |
| Rat Genome 230 2.0 Array | 1392577_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382046_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385216_at | - | - |
| Rat Genome 230 2.0 Array | 1377607_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377607_at | - | - |
| Rat Genome 230 2.0 Array | 1385216_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385814_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382046_at | - | - |
| Rat Genome 230 2.0 Array | 1392577_at | - | - |
| Affymetrix HT Rat Focus Array | 1392577_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377607_at | - | - |
| Affymetrix HT Rat Focus Array | 1382046_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392577_at | - | - |
| Affymetrix HT Rat Focus Array | 1377607_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382046_at | - | - |
| Affymetrix HT Rat Focus Array | 1385814_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385814_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000016287.rn.1 | TC1000001784.rn.1 | RTA TC |
| Rat Expression Set 230 | 1392577_at | - | - |
| Rat Expression Set 230 | 1385814_PM_at | - | - |
| Rat Expression Set 230 | 1392577_PM_at | - | - |
| Rat Expression Set 230 | 1385814_at | - | - |
| Rat Expression Set 230 | 1382046_PM_at | - | - |
| Rat Expression Set 230 | 1377607_PM_at | - | - |
| Rat Expression Set 230 | 1377607_at | - | - |
| Rat Expression Set 230 | 1382046_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739099 | - | - |
| Rat Genome U34 Set | rc_AA925300_at | - | - |
| Rat Genome U34 Set | rc_AA963780_at | - | - |
| Rat Genome U34 Set | rc_AA957862_at | - | - |
| Rat Genome U34 Set | rc_AI179919_at | - | - |
| Rat Genome U34 Set | rc_AA925300_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22152266 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373959 | - | - |

Map3k3
Genomic Map