Assay Details
| Gene Symbol: Tlcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013858.1 | NP_001013880.1 | 1 - 2 | 329 | 77 | |
| GenBank mRNA | BC085876.1 | - | 1 - 2 | 329 | 7191659 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Tlcd1
Genomic Map