Assay Details
| Gene Symbol: Cd300a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205348.1 | NP_001192277.1 | 4 - 5 | 953 | 106 | |
| GenBank mRNA | EF199812.1 | - | 4 - 5 | 648 | 106 | |
| FQ230743.1 | - | 4 - 5 | 718 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd300a
Genomic Map