Assay Details
| Gene Symbol: Lyrm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173471.1 | NP_001166942.1 | 1 - 2 | 51 | 115 | |
| XM_008768097.1 | XP_008766319.1 | 1 - 2 | 53 | 115 | ||
| GenBank mRNA | BC167116.1 | - | 1 - 2 | 51 | 6922274 | 115 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC167116.1, NM_001173471.1, XM_008768097.1 transcripts.
Set Membership: |
|

Lyrm9
Genomic Map