Assay Details
| Gene Symbol: Lyrm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173471.1 | NP_001166942.1 | 2 - 3 | 190 | 64 | |
| GenBank mRNA | BC167116.1 | - | 2 - 3 | 190 | 6922274 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|

Lyrm9
Genomic Map