Assay Details
| Gene Symbol: C1qbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019259.2 | NP_062132.2 | 1 - 2 | 232 | 92 | |
| GenBank mRNA | BC070510.1 | - | 1 - 2 | 232 | 7100505 | 92 |
| FQ212622.1 | - | 1 - 2 | 257 | 92 | ||
| FQ214537.1 | - | 1 - 2 | 282 | 92 | ||
| FQ219796.1 | - | 1 - 2 | 255 | 92 | ||
| FQ220621.1 | - | 1 - 2 | 275 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

C1qbp
Genomic Map