Assay Details
| Gene Symbol: LOC688286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247854.2 | XP_006247916.1 | 1 - 2 | 770 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC688286| Gene Symbol: LOC688286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247854.2 | XP_006247916.1 | 1 - 2 | 770 | 62 | |
Set Membership: |
|