Assay Details
| Gene Symbol: LOC688286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247853.2 | XP_006247915.1 | 4 - 5 | 2210 | 73 | |
| XM_006247854.2 | XP_006247916.1 | 6 - 7 | 1464 | 73 | ||
More Information
Set Membership: |
|

LOC688286
Genomic Map