Assay Details
| Gene Symbol: Slc16a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030834.1 | NP_110461.1 | 2 - 3 | 313 | 59 | |
| XM_006247931.2 | XP_006247993.1 | 3 - 4 | 428 | 59 | ||
| XM_006247932.2 | XP_006247994.1 | 3 - 4 | 720 | 59 | ||
| XM_006247933.2 | XP_006247995.1 | 2 - 3 | 335 | 59 | ||
| XM_006247934.2 | XP_006247996.1 | 3 - 4 | 394 | 59 | ||
| XM_008768513.1 | XP_008766735.1 | 3 - 4 | 3152 | 59 | ||
| XM_008768514.1 | XP_008766736.1 | 2 - 3 | 3059 | 59 | ||
| GenBank mRNA | BC168146.1 | - | 2 - 3 | 272 | 9089640 | 59 |
| U87627.1 | - | 2 - 3 | 313 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc16a3
Genomic Map