Assay Details
| Gene Symbol: Mtfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006960.1 | NP_001006961.1 | 2 - 3 | 322 | 85 | |
| GenBank mRNA | BC083713.1 | - | 2 - 3 | 322 | 7190675 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|

Mtfp1
Genomic Map