Assay Details
| Gene Symbol: Chmp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017466.2 | NP_001017466.2 | 1 - 2 | 300 | 82 | |
| GenBank mRNA | BC092576.1 | - | 1 - 2 | 300 | 7122321 | 82 |
| CO383495.1 | - | 1 - 2 | 278 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chmp4c
Genomic Map